missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,3-benzodioxole, 97%, Maybridge™

Brand:  Maybridge CC01710DA

Code : 31

Additional Details : CAS Number : 6698-13-1 Weight : 0.05100kg

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10009171

  • 338.49 € / 1g

Estimated Shipment
11-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

6698-13-1
201.019
VZPMQHSDFWAZHP-UHFFFAOYSA-N
2776189
C1OC2=C(O1)C(=CC=C2)Br
C7H5BrO2
MFCD02681890
4-bromobenzo d 1,3 dioxole, 4-bromo-2h-1,3-benzodioxole, 4-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 4-bromo, 2,3-methylenedioxy-bromobenzene, 4-bromo-1,3-benzodioxol, methylene dioxy bromo-benzene, 1,3-benzodioxole,4-bromo, 4-bromanyl-1,3-benzodioxole, 1,3-benzodioxol-4-yl bromide
4-bromo-1,3-benzodioxole

Specifications

4-Bromo-1, 3-benzodioxole
1g
96.5
97%
MFCD02681890
VZPMQHSDFWAZHP-UHFFFAOYSA-N
4-bromo-1,3-benzodioxole
2776189
97%
Amber Glass Bottle
6698-13-1
100.0
C7H5BrO2
4-bromobenzo d 1,3 dioxole, 4-bromo-2h-1,3-benzodioxole, 4-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 4-bromo, 2,3-methylenedioxy-bromobenzene, 4-bromo-1,3-benzodioxol, methylene dioxy bromo-benzene, 1,3-benzodioxole,4-bromo, 4-bromanyl-1,3-benzodioxole, 1,3-benzodioxol-4-yl bromide
C1OC2=C(O1)C(=CC=C2)Br
201.019
201.02
Videos
SDS Documents