missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,1,3-benzothiadiazole, 97%, Thermo Scientific™

Catalog Number 10428192
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
22034-13-5
C6H3BrN2S
215.068
MFCD00614355
KYKBVPGDKGABHY-UHFFFAOYSA-N
4-bromobenzo c 1,2,5 thiadiazole, 4-bromo-benzo 1,2,5 thiadiazole, 4-bromobenzo c 1,2,5-thiadiazole, zlchem 1006, 4-bromo-benz-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,4-bromo
736492
4-bromo-2,1,3-benzothiadiazole
C1=CC2=NSN=C2C(=C1)Br
This item is not returnable. View return policy
22034-13-5
C6H3BrN2S
215.068
MFCD00614355
KYKBVPGDKGABHY-UHFFFAOYSA-N
4-bromobenzo c 1,2,5 thiadiazole, 4-bromo-benzo 1,2,5 thiadiazole, 4-bromobenzo c 1,2,5-thiadiazole, zlchem 1006, 4-bromo-benz-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,4-bromo
736492
4-bromo-2,1,3-benzothiadiazole
C1=CC2=NSN=C2C(=C1)Br
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 215.07
Percent Purity 95%
Chemical Name or Material 4-Bromo-2, 1, 3-benzothiadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'