missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N-(4-methoxybenzyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H65921.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15400017

  • 256.00 € / 1g


Estimated Shipment date
from Supplier 21-06-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27975841

Specifications

4-Bromo-2-fluoro-N-(4-methoxybenzyl)benzenesulfonamide
MFCD27975841
Solid
1g
374.22
97%
Videos
SDS