Learn More
4-Bromo-2-fluorobenzyl cyanide, 98+%, ACROS Organics™
Brand: Acros Organics 384450050
Code : 29
Additional Details : CAS Number : 114897-91-5 Weight : 0.05500kg Transport : UN number : 3276 Chem class : 6.1 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
114897-91-5 | |
214.04 | |
QLASQEZPJFNZQC-UHFFFAOYSA-N | |
7172308 | |
C1=CC(=C(C=C1Br)F)CC#N |
C8H5BrFN | |
MFCD07368653 | |
4-bromo-2-fluorobenzyl cyanide, 2-4-bromo-2-fluorophenyl acetonitrile, 4-bromo-2-fluorophenylacetonitrile, 2-fluoro-4-bromophenylacetonitrile, benzeneacetonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorophenyl acetonitrile, 2-4-bromo-2-fluorophenyl ethanenitrile, 4-bromo-2-fluorobenzylcyanide, pubchem10413, acmc-2099lv | |
2-(4-bromo-2-fluorophenyl)acetonitrile |
Specifications
4-Bromo-2-fluorobenzyl cyanide | |
Glass bottle | |
52.8°C to 53.0°C | |
114897-91-5 | |
100.0 | |
C8H5BrFN | |
MFCD07368653 | |
QLASQEZPJFNZQC-UHFFFAOYSA-N | |
2-(4-bromo-2-fluorophenyl)acetonitrile | |
7172308 | |
Crystalline Powder |
Authentic | |
White to Beige | |
5g | |
98.0 | |
98% min. (GC) | |
CNCH2C6H3(F)Br | |
4-bromo-2-fluorobenzyl cyanide, 2-4-bromo-2-fluorophenyl acetonitrile, 4-bromo-2-fluorophenylacetonitrile, 2-fluoro-4-bromophenylacetonitrile, benzeneacetonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorophenyl acetonitrile, 2-4-bromo-2-fluorophenyl ethanenitrile, 4-bromo-2-fluorobenzylcyanide, pubchem10413, acmc-2099lv | |
C1=CC(=C(C=C1Br)F)CC#N | |
214.04 | |
214.04 | |
98+% |
Safety and Handling
- 4-Bromo-2-fluorobenzyl cyanide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.