missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylbenzenesulfonyl chloride, 97%, ACROS Organics™

Brand:  Acros Organics 442390010

Code : 29

Additional Details : CAS Number : 72256-93-0 Weight : 0.05100kg Transport : UN number : 3261 Chem class : 8 Pack group : II


Quantity 1g

 View more versions of this product

Product Code. 11944591

  • 40.59 € / 1g

Estimated Shipment: 06-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

72256-93-0
269.54
JRNDXUKMWFVEIC-UHFFFAOYSA-N
2737503
CC1=C(Br)C=CC(=C1)S(Cl)(=O)=O
C7H6BrClO2S
MFCD03094696
4-bromo-3-methylbenzene-1-sulfonyl chloride, 4-bromo-3-methylbenzenesulphonyl chloride, 4-bromo-3-methylbenzenesulphonylchloride, 4-bromo-3-methyl-benzenesulfonyl chloride, 6-bromo-m-toluene-3-sulfonyl chloride, pubchem15805, acmc-1bho4, jrndxukmwfveic-uhfffaoysa, 4-bromo-3-methylphenyl chlorosulfone, 4-bromo-3-methylbenzene sulfonyl chloride
4-bromo-3-methylbenzenesulfonyl chloride

Specifications

4-Bromo-3-methylbenzenesulfonyl chloride
Glass Bottle
58°C to 60°C
72256-93-0
100.0
C7H6BrClO2S
4-bromo-3-methylbenzene-1-sulfonyl chloride, 4-bromo-3-methylbenzenesulphonyl chloride, 4-bromo-3-methylbenzenesulphonylchloride, 4-bromo-3-methyl-benzenesulfonyl chloride, 6-bromo-m-toluene-3-sulfonyl chloride, pubchem15805, acmc-1bho4, jrndxukmwfveic-uhfffaoysa, 4-bromo-3-methylphenyl chlorosulfone, 4-bromo-3-methylbenzene sulfonyl chloride
CC1=C(Br)C=CC(=C1)S(Cl)(=O)=O
269.54
269.55
97%
Authentic
Brown to White
1g
96.0
96% min. (GC)
MFCD03094696
JRNDXUKMWFVEIC-UHFFFAOYSA-N
4-bromo-3-methylbenzenesulfonyl chloride
2737503
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Bromo-3-methylbenzenesulfonyl chloride

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS Documents
Certificates