missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-3-(trifluoromethyl)-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62138.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15415545

  • 459.78 € / 5g


Estimated Shipment date
from Supplier 29-08-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-3-(trifluoromethyl)-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
MFCD26143308

Specifications

4-Bromo-3-(trifluoromethyl)-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
MFCD26143308
448.18
97%
5g
Solid
Videos
SDS