missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-bromophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62144.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15417086

  • 79.63 € / 1g


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-bromophenyl)-3-(trifluoromethyl)benzenesulfonamide
MFCD26143309

Specifications

4-Bromo-N-(2-bromophenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
97%
459.08
Solid
MFCD26143309
Videos
SDS