missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-bromophenyl)-5-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61180.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416336

  • 85.33 € / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-bromophenyl)-5-fluorobenzenesulfonamide
C12H8Br2FNO2S
409.07
MFCD24848933

Specifications

4-Bromo-N-(2-bromophenyl)-5-fluorobenzenesulfonamide
C12H8Br2FNO2S
MFCD24848933
97%
409.07
1g
Videos
SDS