missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-[4-chloro-3-(trifluoromethyl)phenyl]-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63639.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415635

  • 62.94 € / 1g


Estimated Shipment date
from Supplier 21-11-2018
Add to basket

Specifications

Chemical Identifiers

MFCD26143378

Specifications

4-Bromo-N-[4-chloro-3-(trifluoromethyl)phenyl]-3-(trifluoromethyl)benzenesulfonamide
MFCD26143378
Solid
1g
482.62
97%
Videos
SDS