missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-[4-fluoro-2-(trifluoromethyl)phenyl]-3-(trifluoromethyl)benzenesulfonamide, 97%, Alfa Aesar™

Brand:  Alfa Aesar H63149.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475585

  • 323.00 € / 1g

Estimated Shipment date
from Supplier 18-07-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

MFCD26960688

Specifications

4-Bromo-N-[4-fluoro-2-(trifluoromethyl)phenyl]-3-(trifluoromethyl)benzenesulfonamide
MFCD26960688
Solid
1g
466.17
97%
Videos
SDS