missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-[4-fluoro-3-(trifluoromethyl)phenyl]-3-(trifluoromethyl)benzenesulfonamide, 97%, Alfa Aesar™

Brand:  Alfa Aesar H63230.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475595

  • 323.00 € / 1g


Estimated Shipment date
from Supplier 23-05-2019
Add to basket

Specifications

Chemical Identifiers

MFCD26143355

Specifications

4-Bromo-N-[4-fluoro-3-(trifluoromethyl)phenyl]-3-(trifluoromethyl)benzenesulfonamide
MFCD26143355
Solid
1g
466.17
97%
Videos
SDS