missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzhydrazide, 98+%, Thermo Scientific Chemicals

Catalog Number p-7024374
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
5933-32-4
C7H7BrN2O
215.05
MFCD00007602
UYIMBYKIIMYFPS-UHFFFAOYSA-N
4-bromobenzhydrazide, p-bromobenzhydrazide, 4-bromobenzoic hydrazide, p-bromobenzohydrazide, p-bromobenzoic acid hydrazide, 4-bromobenzoylhydrazine, 4-bromobenzoic acid hydrazide, benzoic acid, 4-bromo-, hydrazide, 4-bromo-benzoic acid hydrazide, p-bromobenzoylhydrazine
22219
4-bromobenzohydrazide
C1=CC(=CC=C1C(=O)NN)Br
This item is not returnable. View return policy
5933-32-4
C7H7BrN2O
215.05
MFCD00007602
UYIMBYKIIMYFPS-UHFFFAOYSA-N
4-bromobenzhydrazide, p-bromobenzhydrazide, 4-bromobenzoic hydrazide, p-bromobenzohydrazide, p-bromobenzoic acid hydrazide, 4-bromobenzoylhydrazine, 4-bromobenzoic acid hydrazide, benzoic acid, 4-bromo-, hydrazide, 4-bromo-benzoic acid hydrazide, p-bromobenzoylhydrazine
22219
4-bromobenzohydrazide
C1=CC(=CC=C1C(=O)NN)Br

4-Bromobenzoic hydrazide has been used in the preparation of {2-[1-oxo-3-(4-bromophenyl)-2-propenyl]hydrazide}-4-bromobenzoic acid, {2,2?-[1,4-phenylenebis(1-oxo-2-propene-3,1-diyl)]dihydrazide}-p-bromobenzoic acid and substituted period[1,2-a]pyrimidine-2,4(3H)-diones (PPMDO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzoic hydrazide has been used in the preparation of {2-[1-oxo-3-(4-bromophenyl)-2-propenyl]hydrazide}-4-bromobenzoic acid, {2,2′-[1,4-phenylenebis(1-oxo-2-propene-3,1-diyl)]dihydrazide}-p-bromobenzoic acid and substituted period[1,2-a]pyrimidine-2,4(3H)-diones (PPMDO).

Solubility
Soluble in hot methanol (almost transparency). Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 167°C to 169°C
Quantity 5 g
Beilstein 777006
Solubility Information Soluble in hot methanol (almost transparency). Slightly soluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzhydrazide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'