missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(2,3-dichlorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59723.03

Code : NEW

Additional Details : CAS Number : 90665-36-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15476118

  • 646.00 € / 1g


Estimated Shipment date
from Supplier 22-05-2019
Add to basket

Specifications

Chemical Identifiers

90665-36-4
300.563
LBYNHHNJVLHJAU-UHFFFAOYSA-N
853841
C1=CC(=C(C(=C1)Cl)Cl)NC(=O)C2=CC=C(C=C2)Cl
C13H8Cl3NO
MFCD00028228
4-chloro-n-2,3-dichlorophenyl benzamide, benzamide, 4-chloro-n-2,3-dichlorophenyl, n-2,3-dichlorophenyl 4-chlorophenyl carboxamide
4-chloro-N-(2,3-dichlorophenyl)benzamide

Specifications

4-Chloro-N-(2,3-dichlorophenyl)benzamide
90665-36-4
MFCD00028228
LBYNHHNJVLHJAU-UHFFFAOYSA-N
4-chloro-N-(2,3-dichlorophenyl)benzamide
853841
Solid
1g
C13H8Cl3NO
4-chloro-n-2,3-dichlorophenyl benzamide, benzamide, 4-chloro-n-2,3-dichlorophenyl, n-2,3-dichlorophenyl 4-chlorophenyl carboxamide
C1=CC(=C(C(=C1)Cl)Cl)NC(=O)C2=CC=C(C=C2)Cl
300.563
300.57
97%
Videos
SDS