missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(2-fluoro-5-methylphenyl)butyramide, 97%

Brand:  Alfa Aesar™ H59737.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415236

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 31-08-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(2-fluoro-5-methylphenyl)butyramide
MFCD18262635

Specifications

4-Chloro-N-(2-fluoro-5-methylphenyl)butyramide
97%
229.68
1g
MFCD18262635
Solid
Videos
SDS