missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3,5-dichlorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59405.03

Code : NEW

Additional Details : CAS Number : 56661-51-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415718

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 03-09-2018
Add to basket

Specifications

Chemical Identifiers

56661-51-9
300.563
JZPOOROGNGIBQQ-UHFFFAOYSA-N
C13H8Cl3NO
MFCD00096063
796539

Specifications

4-Chloro-N-(3,5-dichlorophenyl)benzamide
1g
C13H8Cl3NO
JZPOOROGNGIBQQ-UHFFFAOYSA-N
4-chloro-N-(3,5-dichlorophenyl)benzamide
796539
Solid
56661-51-9
97%
MFCD00096063
C1=CC(=CC=C1C(=O)NC2=CC(=CC(=C2)Cl)Cl)Cl
300.563
300.57
Videos
SDS