missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-ethylbenzamide, 97%, Thermo Scientific™

Catalog Number p-7053107
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
26930-17-6
C9H10ClNO
183.635
MFCD00871156
TXRYLYLDCHDRSJ-UHFFFAOYSA-N
benzamide, 4-chloro-n-ethyl, 4-chloro-n-ethyl-benzamide, n-ethyl-4-chlorobenzamide, 4-chlorobenzamide, n-ethyl, benzamide, p-chloro-n-ethyl, 4-chloro-n-ethylbenzenecarboxamide, 4-chlorophenyl-n-ethylcarboxamide
101292
4-chloro-N-ethylbenzamide
CCNC(=O)C1=CC=C(C=C1)Cl
This item is not returnable. View return policy
26930-17-6
C9H10ClNO
183.635
MFCD00871156
TXRYLYLDCHDRSJ-UHFFFAOYSA-N
benzamide, 4-chloro-n-ethyl, 4-chloro-n-ethyl-benzamide, n-ethyl-4-chlorobenzamide, 4-chlorobenzamide, n-ethyl, benzamide, p-chloro-n-ethyl, 4-chloro-n-ethylbenzenecarboxamide, 4-chlorophenyl-n-ethylcarboxamide
101292
4-chloro-N-ethylbenzamide
CCNC(=O)C1=CC=C(C=C1)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 183.64
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-N-ethylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'