missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals

Catalog Number 11974981
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
122-01-0
C7H4Cl2O
175.01
RKIDDEGICSMIJA-UHFFFAOYSA-N
benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc
8501
CHEBI:60716
4-chlorobenzoyl chloride
C1=CC(=CC=C1C(=O)Cl)Cl
This item is not returnable. View return policy
122-01-0
C7H4Cl2O
175.01
RKIDDEGICSMIJA-UHFFFAOYSA-N
benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc
8501
CHEBI:60716
4-chlorobenzoyl chloride
C1=CC(=CC=C1C(=O)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 12.0°C to 14.0°C
Color Colorless
Density 1.3700g/mL
Boiling Point 222.0°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4COCl
Refractive Index 1.5770 to 1.5790
Quantity 2.5 L
Beilstein 09,341
Specific Gravity 1.37
Solubility Information Solubility in water: hydrolyses. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 175.01
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-Chlorobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'