missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl cyanide, 98+%, Thermo Scientific Chemicals

Catalog Number p-85686
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
140-53-4
C8H6ClN
151.6
MFCD00001918
IVYMIRMKXZAHRV-UHFFFAOYSA-N
4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide
241582
CHEBI:17346
2-(4-chlorophenyl)acetonitrile
C1=CC(=CC=C1CC#N)Cl
This item is not returnable. View return policy
140-53-4
C8H6ClN
151.6
MFCD00001918
IVYMIRMKXZAHRV-UHFFFAOYSA-N
4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide
241582
CHEBI:17346
2-(4-chlorophenyl)acetonitrile
C1=CC(=CC=C1CC#N)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 29°C to 30°C
Color Colorless to Yellow
Boiling Point 265°C to 267°C
Flash Point 133°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CH2CN
Refractive Index 1.5415 to 1.5435
Quantity 5 g
Beilstein 09, 448
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone and alcohol
Formula Weight 151.6
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 4-Chlorobenzyl cyanide, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorobenzyl cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'