missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylhydrazine Hydrochloride 97%, ACROS Organics™

Brand:  Acros Organics 404950050

Code : 29

Additional Details : CAS Number : 1073-70-7 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 10425872

  • 22.20 € / 5g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

1073-70-7
179.044
YQVZREHUWCCHHX-UHFFFAOYSA-N
C6H8Cl2N2
MFCD00012943
71600

Specifications

4-Chlorophenylhydrazine hydrochloride
203.0°C
97%
96.0
Authentic
Glass bottle
97%
NHNH2C6H4Cl·HCl
YQVZREHUWCCHHX-UHFFFAOYSA-N
(4-chlorophenyl)hydrazine;hydrochloride
71600
Crystalline Powder
Pink to White
5g
1073-70-7
100.0
1% max.
97%
C6H8Cl2N2
MFCD00012943
C1=CC(=CC=C1NN)Cl.Cl
179.044
179.05
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Chlorophenylhydrazine hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS