missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylacetic acid, 97%, ACROS Organics™

Brand:  Acros Organics 437730050

Code : 29

Additional Details : CAS Number : 5462-71-5 Weight : 0.05500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 11945631

  • 363.00 € / 5g

Estimated Shipment: 05-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

5462-71-5
161.16
WEBXRQONNWEETE-UHFFFAOYSA-N
79587
OC(=O)CC1=CC=C(C=C1)C#N
C9H7NO2
MFCD06798066
2-4-cyanophenyl acetic acid, 4-cyanophenylacetic acid, 4-cyanophenyl acetic acid, benzeneacetic acid, 4-cyano, 4-cyano-phenyl-acetic acid, p-cyanophenylacetic acid, 4-cyanobenzeneacetic acid, p-cyanophenyl acetic acid, acetic acid, p-cyanophenyl
2-(4-cyanophenyl)acetic acid

Specifications

4-Cyanophenylacetic acid
Glass bottle
151°C to 152°C
5462-71-5
100.0
C9H7NO2
2-4-cyanophenyl acetic acid, 4-cyanophenylacetic acid, 4-cyanophenyl acetic acid, benzeneacetic acid, 4-cyano, 4-cyano-phenyl-acetic acid, p-cyanophenylacetic acid, 4-cyanobenzeneacetic acid, p-cyanophenyl acetic acid, acetic acid, p-cyanophenyl
OC(=O)CC1=CC=C(C=C1)C#N
161.16
161.16
97%
Authentic
Brown to White
5g
96.0
96% min. (HPLC)
MFCD06798066
WEBXRQONNWEETE-UHFFFAOYSA-N
2-(4-cyanophenyl)acetic acid
79587
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Cyanophenylacetic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates