missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog Number 11935631
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5462-71-5
C9H7NO2
161.16
MFCD06798066
WEBXRQONNWEETE-UHFFFAOYSA-N
2-4-cyanophenyl acetic acid, 4-cyanophenylacetic acid, 4-cyanophenyl acetic acid, benzeneacetic acid, 4-cyano, 4-cyano-phenyl-acetic acid, p-cyanophenylacetic acid, 4-cyanobenzeneacetic acid, p-cyanophenyl acetic acid, acetic acid, p-cyanophenyl
79587
2-(4-cyanophenyl)acetic acid
OC(=O)CC1=CC=C(C=C1)C#N
This item is not returnable. View return policy
5462-71-5
C9H7NO2
161.16
MFCD06798066
WEBXRQONNWEETE-UHFFFAOYSA-N
2-4-cyanophenyl acetic acid, 4-cyanophenylacetic acid, 4-cyanophenyl acetic acid, benzeneacetic acid, 4-cyano, 4-cyano-phenyl-acetic acid, p-cyanophenylacetic acid, 4-cyanobenzeneacetic acid, p-cyanophenyl acetic acid, acetic acid, p-cyanophenyl
79587
2-(4-cyanophenyl)acetic acid
OC(=O)CC1=CC=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 151°C to 152°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 161.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Cyanophenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Cyanophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'