missing translation for 'onlineSavingsMsg'
Learn More

4-(Difluoromethoxy)phenyl isocyanate, 99%, Thermo Scientific™

Catalog Number p-4089992
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
58417-15-5
C8H5F2NO2
185.13
MFCD00673056
XLUSNOUGZQJPRN-UHFFFAOYSA-N
4-difluoromethoxy phenyl isocyanate, 1-difluoromethoxy-4-isocyanatobenzene, 4-difluoromethoxy benzenisocyanate, acmc-1b0ih, 4-difluoromethoxyphenyl isocyanate, 4-difluoromethoxy-phenyl isocyanate, 4-difluoromethoxy phenyl isosyanate, 1-difluoromethoxy-4-isocyanato-benzene, 4-difluoromethoxy phenylisocyanate, benzene,1-difluoromethoxy-4-isocyanato
2733371
1-(difluoromethoxy)-4-isocyanatobenzene
FC(F)OC1=CC=C(C=C1)N=C=O
This item is not returnable. View return policy
58417-15-5
C8H5F2NO2
185.13
MFCD00673056
XLUSNOUGZQJPRN-UHFFFAOYSA-N
4-difluoromethoxy phenyl isocyanate, 1-difluoromethoxy-4-isocyanatobenzene, 4-difluoromethoxy benzenisocyanate, acmc-1b0ih, 4-difluoromethoxyphenyl isocyanate, 4-difluoromethoxy-phenyl isocyanate, 4-difluoromethoxy phenyl isosyanate, 1-difluoromethoxy-4-isocyanato-benzene, 4-difluoromethoxy phenylisocyanate, benzene,1-difluoromethoxy-4-isocyanato
2733371
1-(difluoromethoxy)-4-isocyanatobenzene
FC(F)OC1=CC=C(C=C1)N=C=O
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.3230g/mL
Boiling Point 209.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula F2CHOC6H4NCO
Refractive Index 1.4940 to 1.4960
Quantity 500 mg
Specific Gravity 1.323
Formula Weight 185.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-(Difluoromethoxy)phenyl isocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-(Difluoromethoxy)phenyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'