Learn More
4'-Ethoxyacetophenone, 99%, ACROS Organics™
Brand: Acros Organics 409595000
Code : 29
Additional Details : CAS Number : 1676-63-7 Weight : 0.55000kg
Quantity | 500g |
---|
Chemical Identifiers
1676-63-7 | |
164.2 | |
YJFNFQHMQJCPRG-UHFFFAOYSA-N | |
72872 | |
CCOC1=CC=C(C=C1)C(=O)C |
C10H12O2 | |
MFCD00009095 | |
4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone | |
1-(4-ethoxyphenyl)ethanone |
Specifications
4'-Ethoxyacetophenone | |
268.0°C to 269.0°C (758.0mmHg) | |
500g | |
98.5 | |
98.5% min. (GC) | |
MFCD00009095 | |
YJFNFQHMQJCPRG-UHFFFAOYSA-N | |
1-(4-ethoxyphenyl)ethanone | |
72872 | |
99% |
134°C | |
35.0°C to 38.0°C | |
1676-63-7 | |
100.0 | |
C10H12O2 | |
4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone | |
CCOC1=CC=C(C=C1)C(=O)C | |
164.2 | |
164.2 |
Safety and Handling
- 4'-Ethoxyacetophenone
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.