missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-4228641
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
16331-46-7
C9H9ClO2
184.62
XLWQUESMILVIPR-UHFFFAOYSA-N
4-ethoxybenzoylchloride, p-ethoxybenzoyl chloride, p-ethoxy benzoyl chloride, 4-ethoxy-benzoyl chloride, benzoyl chloride, 4-ethoxy, pubchem10868, acmc-209dpj, 4-ethyloxybenzoyl chloride, para-ethoxybenzoyl chloride, ksc493i8n
140059
4-ethoxybenzoyl chloride
CCOC1=CC=C(C=C1)C(=O)Cl
This item is not returnable. View return policy
16331-46-7
C9H9ClO2
184.62
XLWQUESMILVIPR-UHFFFAOYSA-N
4-ethoxybenzoylchloride, p-ethoxybenzoyl chloride, p-ethoxy benzoyl chloride, 4-ethoxy-benzoyl chloride, benzoyl chloride, 4-ethoxy, pubchem10868, acmc-209dpj, 4-ethyloxybenzoyl chloride, para-ethoxybenzoyl chloride, ksc493i8n
140059
4-ethoxybenzoyl chloride
CCOC1=CC=C(C=C1)C(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow or White to Yellow
Boiling Point 128°C to 130°C (6.0 Torr)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 184.62
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Ethoxybenzoyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Ethoxybenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'