missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethynylbenzonitrile, 97%, Thermo Scientific Chemicals
54.46 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
4-Ethynylbenzonitrile is used as a synthetic fragment and as a test compound for cross-coupling reactions. It is used in the study of hydrogen bond formation in multifunctional molecules due to the presence of four hydrogen bonding sites. It is also involved in the preparation of 4-[(trimethylsilyl)ethynyl]benzonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Ethynylbenzonitrile is used as a synthetic fragment and as a test compound for cross-coupling reactions. It is used in the study of hydrogen bond formation in multifunctional molecules due to the presence of four hydrogen bonding sites. It is also involved in the preparation of 4-[(trimethylsilyl)ethynyl]benzonitrile.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents, acids and organic materials.
4-Ethynylbenzonitrile is used as a synthetic fragment and as a test compound for cross-coupling reactions. It is used in the study of hydrogen bond formation in multifunctional molecules due to the presence of four hydrogen bonding sites. It is also involved in the preparation of 4-[(trimethylsilyl)ethynyl]benzonitrile.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents, acids and organic materials.
Specifications
Melting Point | 156°C to 160°C |
Color | Yellow |
Quantity | 1 g |
UN Number | UN3439 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 127.14 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 4-Ethynylbenzonitrile |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'