missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrotoluene, 98%, ACROS Organics™

Brand:  Acros Organics 119520100

Code : 29

Additional Details : CAS Number : 446-10-6 Weight : 0.06000kg


Packaging Glass bottle
Quantity 10g

 View more versions of this product

Product Code. 10001090

  • 26.20 € / 10g

Estimated Shipment
24-04-2020

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

446-10-6
155.13
SKWTUNAAJNDEIK-UHFFFAOYSA-N
67965
CC1=C(C=C(C=C1)F)[N+](=O)[O-]
C7H6FNO2
MFCD00007200
4-fluoro-2-nitrotoluene, 2-nitro-4-fluorotoluene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, 4-fluoro-2-nitrobenzene, 5-fluoro-2-methylnitrobenzene, 4-fluoro-2-nitro toluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoronitrotoluene, zlchem 482
4-fluoro-1-methyl-2-nitrobenzene

Specifications

4-Fluoro-2-nitrotoluene
Authentic
1.5218 to 1.5238
Amber to Yellow
10g
446-10-6
100.0
C7H6FNO2
MFCD00007200
4-fluoro-2-nitrotoluene, 2-nitro-4-fluorotoluene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, 4-fluoro-2-nitrobenzene, 5-fluoro-2-methylnitrobenzene, 4-fluoro-2-nitro toluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoronitrotoluene, zlchem 482
CC1=C(C=C(C=C1)F)[N+](=O)[O-]
155.13
155.13
98%
98°C
Glass bottle
138°C to 139°C (83.0mmHg)
27°C
98%
97.5
97.5% min. (GC)
CH3C6H3(NO2)F
05, I, 161
SKWTUNAAJNDEIK-UHFFFAOYSA-N
4-fluoro-1-methyl-2-nitrobenzene
67965
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Fluoro-2-nitrotoluene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents