missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-formylphenylboronic acid, 95%, Thermo Scientific™

Catalog Number p-4380069
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
374538-01-9
C7H6BFO3
167.93
YABSTJQEBSKPCG-UHFFFAOYSA-N
4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, boronic acid, 4-fluoro-3-formylphenyl, 4-fluoro-3-formyl-phenyl boronic acid, pubchem10809, acmc-1cswy, 3-formyl 4-fluorophenyl boronic acid, 5-borono-2-fluorobenzaldehyde, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid
2778651
(4-fluoro-3-formylphenyl)boronic acid
B(C1=CC(=C(C=C1)F)C=O)(O)O
This item is not returnable. View return policy
374538-01-9
C7H6BFO3
167.93
YABSTJQEBSKPCG-UHFFFAOYSA-N
4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, boronic acid, 4-fluoro-3-formylphenyl, 4-fluoro-3-formyl-phenyl boronic acid, pubchem10809, acmc-1cswy, 3-formyl 4-fluorophenyl boronic acid, 5-borono-2-fluorobenzaldehyde, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid
2778651
(4-fluoro-3-formylphenyl)boronic acid
B(C1=CC(=C(C=C1)F)C=O)(O)O
Melting Point 132°C to 134°C
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 167.93
Percent Purity 95%
Chemical Name or Material 4-Fluoro-3-formylphenylboronic acid
compliance-icons
Product Identifier
  • 4-Fluoro-3-formylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'