missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-formylphenylboronic acid, 95%, ACROS Organics™

Brand:  Acros Organics 445440050

Code : 29

Additional Details : CAS Number : 374538-01-9 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11966141

  • 146.00 € / 5g

Estimated Shipment
29-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

374538-01-9
167.93
4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, boronic acid, 4-fluoro-3-formylphenyl, 4-fluoro-3-formyl-phenyl boronic acid, pubchem10809, acmc-1cswy, 3-formyl 4-fluorophenyl boronic acid, 5-borono-2-fluorobenzaldehyde, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid
(4-fluoro-3-formylphenyl)boronic acid
C7H6BFO3
YABSTJQEBSKPCG-UHFFFAOYSA-N
2778651
B(C1=CC(=C(C=C1)F)C=O)(O)O

Specifications

4-Fluoro-3-formylphenylboronic acid
5g
94% min. (HPLC)
4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, boronic acid, 4-fluoro-3-formylphenyl, 4-fluoro-3-formyl-phenyl boronic acid, pubchem10809, acmc-1cswy, 3-formyl 4-fluorophenyl boronic acid, 5-borono-2-fluorobenzaldehyde, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid
B(C1=CC(=C(C=C1)F)C=O)(O)O
167.93
167.93
132°C to 134°C
374538-01-9
C7H6BFO3
YABSTJQEBSKPCG-UHFFFAOYSA-N
(4-fluoro-3-formylphenyl)boronic acid
2778651
95%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Fluoro-3-formylphenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents