Learn More
4'-Formyl(1,1'-biphenyl)-4-carboxylic acid, 97%, Maybridge™
Brand: Maybridge MO01260DA
Code : 31
Additional Details : CAS Number : 70916-98-2 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
70916-98-2 | |
226.231 | |
JCEAFZUEUSBESW-UHFFFAOYSA-N | |
2794722 | |
C1=CC(=CC=C1C=O)C2=CC=C(C=C2)C(=O)O |
C14H10O3 | |
MFCD03424613 | |
4-4-formylphenyl benzoic acid, 4'-formylbiphenyl-4-carboxylic acid, 4'-formyl-1,1'-biphenyl-4-carboxylic acid, 4'-formyl-biphenyl-4-carboxylic acid, 4-biphenyl-4'-formyl-carboxylic acid, 4'-formyl 1,1'-biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, 4'-formyl, 4-biphenyl-4'-formyl-carboxylicacid, pubchem10282 | |
4-(4-formylphenyl)benzoic acid |
Specifications
4'-Formyl(1, 1'-biphenyl)-4-carboxylic acid | |
1g | |
97.0 | |
C14H10O3 | |
4-4-formylphenyl benzoic acid, 4'-formylbiphenyl-4-carboxylic acid, 4'-formyl-1,1'-biphenyl-4-carboxylic acid, 4'-formyl-biphenyl-4-carboxylic acid, 4-biphenyl-4'-formyl-carboxylic acid, 4'-formyl 1,1'-biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, 4'-formyl, 4-biphenyl-4'-formyl-carboxylicacid, pubchem10282 | |
C1=CC(=CC=C1C=O)C2=CC=C(C=C2)C(=O)O | |
226.231 | |
226.23 |
Amber glass bottle | |
70916-98-2 | |
97% | |
MFCD03424613 | |
JCEAFZUEUSBESW-UHFFFAOYSA-N | |
4-(4-formylphenyl)benzoic acid | |
2794722 | |
97% |