missing translation for 'onlineSavingsMsg'
Learn More

4-Hexen-1-ol, 97%, predominantly trans, Thermo Scientific Chemicals

Catalog Number 11954621
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
928-92-7
C6H12O
100.16
MFCD00009713
VTIODUHBZHNXFP-NSCUHMNNSA-N
4-hexen-1-ol, e-4-hexen-1-ol, e-hex-4-en-1-ol, trans-4-hexen-1-ol, 4-hexen-1-ol, 4e, 4-hexen-1-ol, e, trans-4-hexenol, unii-zhb2532c2h, 4-hexen-1-ol, predominantly trans
641248
C\C=C\CCCO
This item is not returnable. View return policy
928-92-7
C6H12O
100.16
MFCD00009713
VTIODUHBZHNXFP-NSCUHMNNSA-N
4-hexen-1-ol, e-4-hexen-1-ol, e-hex-4-en-1-ol, trans-4-hexen-1-ol, 4-hexen-1-ol, 4e, 4-hexen-1-ol, e, trans-4-hexenol, unii-zhb2532c2h, 4-hexen-1-ol, predominantly trans
641248
C\C=C\CCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8500g/mL
Boiling Point 159.0°C to 160.0°C
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CH(CH2)3OH
Refractive Index 1.4394
Quantity 5 g
Beilstein 01,III,1929
Specific Gravity 0.85
IUPAC Name (E)-hex-4-en-1-ol
Formula Weight 100.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Hexen-1-ol
Show More Show Less
Product Identifier
  • 4-Hexen-1-ol, predominantly trans

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'