missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzaldehyde, 99%, ACROS Organics™

Brand:  Acros Organics 162770025

Code : 29

Additional Details : CAS Number : 123-08-0 Weight : 2.50000kg


Packaging Plastic bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 11954791

  • 516.00 € / 2.50kg

Estimated Shipment
06-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

123-08-0
122.12
p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde
CHEBI:17597
C1=CC(=CC=C1C=O)O
C7H6O2
RGHHSNMVTDWUBI-UHFFFAOYSA-N
126
4-hydroxybenzaldehyde

Specifications

4-Hydroxybenzaldehyde
Plastic bottle
114.0°C to 118.0°C
123-08-0
C7H6O2
08,64
p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde
RGHHSNMVTDWUBI-UHFFFAOYSA-N
4-hydroxybenzaldehyde
126
122.12
99%
Authentic
Yellow to Brown
2.5kg
98.5% min. (GC)
HOC6H4CHO
15,4849
Solubility in water: 13g/L (30°C). Other solubilities: 36.8g/L benzene (65°C)
C1=CC(=CC=C1C=O)O
122.12
CHEBI:17597
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Hydroxybenzaldehyde

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS Documents