missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxybenzenesulfonic acid, 65%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
Description
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Density | 1.337 g/mL |
Refractive Index | 1.489 |
Quantity | 1 L |
UN Number | UN1803 |
Beilstein | 1869034 |
Solubility Information | Soluble in water. |
Formula Weight | 174.17 |
Percent Purity | 65% |
Physical Form | Liquid |
Chemical Name or Material | 4-Hydroxybenzenesulfonic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'