missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-7022361
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
1137-42-4
C13H10O2
198.22
MFCD00002355
NPFYZDNDJHZQKY-UHFFFAOYSA-N
4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone
14347
CHEBI:34421
(4-hydroxyphenyl)-phenylmethanone
OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
This item is not returnable. View return policy
1137-42-4
C13H10O2
198.22
MFCD00002355
NPFYZDNDJHZQKY-UHFFFAOYSA-N
4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone
14347
CHEBI:34421
(4-hydroxyphenyl)-phenylmethanone
OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 132°C to 135°C
Boiling Point 260°C to 262°C (24 mmHg)
Quantity 50 g
Beilstein 777776
Solubility Information Insoluble in water.
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 4-Hydroxybenzophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'