missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzylamine hydrate, 97%, Thermo Scientific Chemicals

Catalog Number p-4228717
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
696-60-6
C7H9NO
123.16
MFCD19690971
RQJDUEKERVZLLU-UHFFFAOYSA-N
4-hydroxybenzylamine, 4-aminomethyl phenol, 4-aminomethyl-phenol, para-hydroxybenzylamine, unii-7j7f85b7bi, 4-hydroxyphenyl methanaminium bromide, a-amino-p-cresol, 4-hyroxybenzylamine, p-hydroxybenzylamine, 4-hydroxylbenzylamine
97472
4-(aminomethyl)phenol
NCC1=CC=C(O)C=C1
This item is not returnable. View return policy
696-60-6
C7H9NO
123.16
MFCD19690971
RQJDUEKERVZLLU-UHFFFAOYSA-N
4-hydroxybenzylamine, 4-aminomethyl phenol, 4-aminomethyl-phenol, para-hydroxybenzylamine, unii-7j7f85b7bi, 4-hydroxyphenyl methanaminium bromide, a-amino-p-cresol, 4-hyroxybenzylamine, p-hydroxybenzylamine, 4-hydroxylbenzylamine
97472
4-(aminomethyl)phenol
NCC1=CC=C(O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 100°C to 104°C
Color White to Yellow
Elemental Analysis 57.18 to 61.94% C, 9.52 to 10.31% N (based on monohydrate)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 141.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Hydroxybenzylamine hydrate
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hydroxybenzylamine hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'