Learn More
4-Hydroxymethylbenzoic acid, 98%, ACROS Organics™
Brand: Acros Organics 394340010
Code : 29
Additional Details : CAS Number : 3006-96-0 Weight : 0.05100kg
Packaging | Glass bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
3006-96-0 | |
152.15 | |
4-hydroxymethyl benzoic acid, 4-hydroxythylbenzoic acid, unii-9z28l8sthd, hmba linker, 4-hydroxymethyl-benzoic acid, benzoic acid, 4-hydroxymethyl, p-hydroxymethyl benzoic acid, 4-hydroxymethyl benzoicacid, 9z28l8sthd | |
4-(hydroxymethyl)benzoic acid |
C8H8O3 | |
WWYFPDXEIFBNKE-UHFFFAOYSA-N | |
76360 | |
C1=CC(=CC=C1CO)C(=O)O |
Specifications
4-Hydroxymethylbenzoic acid | |
Glass bottle | |
182.0°C to 184.0°C | |
3006-96-0 | |
100.0 | |
C8H8O3 | |
4-hydroxymethyl benzoic acid, 4-hydroxythylbenzoic acid, unii-9z28l8sthd, hmba linker, 4-hydroxymethyl-benzoic acid, benzoic acid, 4-hydroxymethyl, p-hydroxymethyl benzoic acid, 4-hydroxymethyl benzoicacid, 9z28l8sthd | |
C1=CC(=CC=C1CO)C(=O)O | |
152.15 | |
152.15 | |
98% |
Authentic | |
White | |
1g | |
97.5 | |
97.5% min. (HPLC) | |
COOHC6H4CH2OH | |
WWYFPDXEIFBNKE-UHFFFAOYSA-N | |
4-(hydroxymethyl)benzoic acid | |
76360 | |
Coarse Fibers |
Safety and Handling
- 4-Hydroxymethylbenzoic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.