Learn More
4-Iodophenyl isothiocyanate, 97%, Maybridge™
Brand: Maybridge S00350DA
Code : 31
Additional Details : CAS Number : 2059-76-9 Weight : 0.05100kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
2059-76-9 | |
261.08 | |
SNLGBRZZFRSXHA-UHFFFAOYSA-N | |
74938 | |
C1=CC(=CC=C1N=C=S)I |
C7H4INS | |
MFCD00041091 | |
4-iodophenyl isothiocyanate, 4-iodophenylisothiocyanate, 1-iodo-4-isothiocyanato-benzene, benzene, 1-iodo-4-isothiocyanato, 4-ipitc, acmc-20amg6, 4-iodobenzenisothiocyanate, 1-isothiocyanato-4-iodobenzene, #, benzene,1-iodo-4-isothiocyanato | |
1-iodo-4-isothiocyanatobenzene |
Specifications
4-Iodophenyl isothiocyanate | |
1g | |
97.0 | |
C7H4INS | |
MFCD00041091 | |
SNLGBRZZFRSXHA-UHFFFAOYSA-N | |
1-iodo-4-isothiocyanatobenzene | |
74938 | |
97% |
Amber Glass Bottle | |
2059-76-9 | |
97% | |
IC6H4NCS | |
4-iodophenyl isothiocyanate, 4-iodophenylisothiocyanate, 1-iodo-4-isothiocyanato-benzene, benzene, 1-iodo-4-isothiocyanato, 4-ipitc, acmc-20amg6, 4-iodobenzenisothiocyanate, 1-isothiocyanato-4-iodobenzene, #, benzene,1-iodo-4-isothiocyanato | |
C1=CC(=CC=C1N=C=S)I | |
261.08 | |
261.08 |
Safety and Handling
- 4-Iodophenyl isothiocyanate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.