missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenyl isothiocyanate, 97%, Maybridge™

Brand:  Maybridge S00350EA

Code : 31

Additional Details : CAS Number : 2059-76-9 Weight : 0.06000kg


Packaging Amber Glass Bottle
Quantity 10g

 View more versions of this product

Product Code. 10009601

  • 35.55 € / 10g

Estimated Shipment: 24-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

2059-76-9
261.08
SNLGBRZZFRSXHA-UHFFFAOYSA-N
74938
IC1=CC=C(C=C1)N=C=S
C7H4INS
MFCD00041091
4-iodophenyl isothiocyanate, 4-iodophenylisothiocyanate, 1-iodo-4-isothiocyanato-benzene, benzene, 1-iodo-4-isothiocyanato, 4-ipitc, acmc-20amg6, 4-iodobenzenisothiocyanate, 1-isothiocyanato-4-iodobenzene, #, benzene,1-iodo-4-isothiocyanato
1-iodo-4-isothiocyanatobenzene

Specifications

4-Iodophenyl isothiocyanate
10g
97.0
C7H4INS
MFCD00041091
SNLGBRZZFRSXHA-UHFFFAOYSA-N
1-iodo-4-isothiocyanatobenzene
74938
97%
Amber Glass Bottle
2059-76-9
97%
IC6H4NCS
4-iodophenyl isothiocyanate, 4-iodophenylisothiocyanate, 1-iodo-4-isothiocyanato-benzene, benzene, 1-iodo-4-isothiocyanato, 4-ipitc, acmc-20amg6, 4-iodobenzenisothiocyanate, 1-isothiocyanato-4-iodobenzene, #, benzene,1-iodo-4-isothiocyanato
IC1=CC=C(C=C1)N=C=S
261.08
261.08
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Iodophenyl isothiocyanate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents
Certificates