missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methoxy-N-(3-methylbenzyl)benzamide, 97%

Brand:  Alfa Aesar™ H59560.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415928

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Methoxy-N-(3-methylbenzyl)benzamide
255.32
MFCD04068178

Specifications

4-Methoxy-N-(3-methylbenzyl)benzamide
Solid
255.32
1g
97%
MFCD04068178
Videos
SDS