missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methoxy-N-(3-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59802.03

Code : NEW

Additional Details : CAS Number : 7464-53-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416218

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 03-09-2018
Add to basket

Specifications

Chemical Identifiers

7464-53-1
241.29
DOEUHYAIQRIZIY-UHFFFAOYSA-N
C15H15NO2
MFCD00442785
346046

Specifications

4-Methoxy-N-(3-methylphenyl)benzamide
7464-53-1
C15H15NO2
DOEUHYAIQRIZIY-UHFFFAOYSA-N
4-methoxy-N-(3-methylphenyl)benzamide
346046
Solid
1g
97%
MFCD00442785
CC1=CC(=CC=C1)NC(=O)C2=CC=C(C=C2)OC
241.29
241.29
Videos
SDS