missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxychalcone, 98%, Thermo Scientific™

Catalog Number p-2371110
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
959-33-1
C16H14O2
238.29
MFCD00017179
XUFXKBJMCRJATM-FMIVXFBMSA-N
4-methoxychalcone, e-3-4-methoxyphenyl-1-phenylprop-2-en-1-one, 2e-3-4-methoxyphenyl-1-phenylprop-2-en-1-one, chalcone, 4-methoxy, 3-4-methoxyphenyl-1-phenyl-2-propen-1-one, 2-propen-1-one, 3-4-methoxyphenyl-1-phenyl, 2-propen-1-one, 3-4-methoxyphenyl-1-phenyl-, 2e, phenyl p-methoxystyryl ketone, 4'-methoxybenzylideneacetophenone, 3-4-methoxyphenyl-1-phenylprop-2-en-1-one
641819
(E)-3-(4-methoxyphenyl)-1-phenylprop-2-en-1-one
COC1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2
This item is not returnable. View return policy
959-33-1
C16H14O2
238.29
MFCD00017179
XUFXKBJMCRJATM-FMIVXFBMSA-N
4-methoxychalcone, e-3-4-methoxyphenyl-1-phenylprop-2-en-1-one, 2e-3-4-methoxyphenyl-1-phenylprop-2-en-1-one, chalcone, 4-methoxy, 3-4-methoxyphenyl-1-phenyl-2-propen-1-one, 2-propen-1-one, 3-4-methoxyphenyl-1-phenyl, 2-propen-1-one, 3-4-methoxyphenyl-1-phenyl-, 2e, phenyl p-methoxystyryl ketone, 4'-methoxybenzylideneacetophenone, 3-4-methoxyphenyl-1-phenylprop-2-en-1-one
641819
(E)-3-(4-methoxyphenyl)-1-phenylprop-2-en-1-one
COC1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 73°C to 78°C
Color Yellow
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dichloromethane,methanol
Formula Weight 238.29
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxychalcone
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxychalcone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'