missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetone, 98+%, Thermo Scientific Chemicals

Catalog Number p-158653
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
122-84-9
C10H12O2
164.2
MFCD00008773
WFWKNGZODAOLEO-UHFFFAOYSA-N
4-methoxyphenylacetone, p-acetonylanisole, anisketone, 1-4-methoxyphenyl propan-2-one, 2-propanone, 1-4-methoxyphenyl, anisyl methyl ketone, p-methoxyphenylacetone, 4-methoxybenzyl methyl ketone, 1-p-methoxyphenyl-2-propanone, anisic ketone
31231
1-(4-methoxyphenyl)propan-2-one
CC(=O)CC1=CC=C(C=C1)OC
This item is not returnable. View return policy
122-84-9
C10H12O2
164.2
MFCD00008773
WFWKNGZODAOLEO-UHFFFAOYSA-N
4-methoxyphenylacetone, p-acetonylanisole, anisketone, 1-4-methoxyphenyl propan-2-one, 2-propanone, 1-4-methoxyphenyl, anisyl methyl ketone, p-methoxyphenylacetone, 4-methoxybenzyl methyl ketone, 1-p-methoxyphenyl-2-propanone, anisic ketone
31231
1-(4-methoxyphenyl)propan-2-one
CC(=O)CC1=CC=C(C=C1)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Green-Yellow to Yellow
Density 1.0670g/mL
Boiling Point 266°C to 268°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4CH2COCH3
Refractive Index 1.5200 to 1.53
Quantity 5 g
Beilstein 08, IV, 460
Specific Gravity 1.067
Formula Weight 164.2
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenylacetone, 98+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'