Learn More
4-Methoxyphenylmagnesium bromide, 1M solution in THF, AcroSeal™, ACROS Organics™
Brand: Acros Organics 377421000
Code : 29
Additional Details : CAS Number : 13139-86-1 Weight : 0.15000kg Transport : UN number : 2924 Chem class : 3 Pack group : II
Packaging | AcroSeal™ Glass bottle |
---|---|
Quantity | 100mL |
Chemical Identifiers
13139-86-1 | |
211.34 | |
RBWRWAUAVRMBAC-UHFFFAOYSA-M | |
2734898 | |
COC1=CC=[C-]C=C1.[Mg+2].[Br-] |
C7H7BrMgO | |
MFCD00013515 | |
4-methoxyphenylmagnesium bromide, grignard reagent, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide | |
magnesium;methoxybenzene;bromide |
Specifications
4-Methoxyphenylmagnesium bromide | |
−21°C | |
1 | |
100mL | |
74.64 | |
C7H7BrMgO | |
MFCD00013515 | |
Solubility in water: vigorous reaction | |
COC1=CC=[C-]C=C1.[Mg+2].[Br-] | |
211.34 | |
211.34 |
1.0000g/mL | |
AcroSeal™ Glass bottle | |
Yellow to Brown | |
13139-86-1,109-99-9 | |
80.98 | |
CH3O(C6H4)MgBr | |
4-methoxyphenylmagnesium bromide, grignard reagent, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide | |
RBWRWAUAVRMBAC-UHFFFAOYSA-M | |
magnesium;methoxybenzene;bromide | |
2734898 | |
Liquid |
Safety and Handling
- 4-Methoxyphenylmagnesium bromide, 1M solution in THF
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.