missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2,1,3-benzoxadiazole, 95%, Thermo Scientific™

Catalog Number p-4259010
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
29091-40-5
C7H6N2O
134.14
MFCD00100600
PTXJEYIAUIOTSH-UHFFFAOYSA-N
4-methylbenzo c 1,2,5 oxadiazole, 4-methylbenzofurazan, 4-methyl-benzofurazan, 4-methyl-benzo 1,2,5 oxadiazole, 2,1,3-benzoxadiazole, 4-methyl, methylbenzfurazan, 2,1,3-benzoxadiazole,4-methyl, #
584114
4-methyl-2,1,3-benzoxadiazole
CC1=CC=CC2=NON=C12
This item is not returnable. View return policy
29091-40-5
C7H6N2O
134.14
MFCD00100600
PTXJEYIAUIOTSH-UHFFFAOYSA-N
4-methylbenzo c 1,2,5 oxadiazole, 4-methylbenzofurazan, 4-methyl-benzofurazan, 4-methyl-benzo 1,2,5 oxadiazole, 2,1,3-benzoxadiazole, 4-methyl, methylbenzfurazan, 2,1,3-benzoxadiazole,4-methyl, #
584114
4-methyl-2,1,3-benzoxadiazole
CC1=CC=CC2=NON=C12
CAS Min % 94.0
CAS Max % 100.0
Melting Point 40.0°C to 41.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 134.14
Percent Purity 95%
Physical Form Crystalline Powder or Low Melting Solid
Chemical Name or Material 4-Methyl-2, 1, 3-benzoxadiazole
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methyl-2,1,3-benzoxadiazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'