missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentanone, 99.5%, for HPLC, Thermo Scientific Chemicals
47.40 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
Specifications
Alkalinity | 0.0001 meq/g max. |
Acidity | 0.0001 meq/g max. |
CAS | 108-10-1 |
Absorbance | 0.01 max. at 400nm, 0.02 max. at 380nm, 0.15 max. at 360nm, 0.5 max. at 340nm, 1 max. at 335nm |
CAS Min % | 99.4 |
CAS Max % | 100.0 |
Melting Point | -84.0°C |
Density | 0.8000g/mL |
Boiling Point | 117.4°C |
Flash Point | 14°C |
Show More |
Safety and Handling
Product Identifier
- 4-Methyl-2-pentanone
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 2
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'