Learn More
4-Methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole, 97%, Maybridge™
Brand: Maybridge CC29239CB
Code : 31
Additional Details : CAS Number : 690632-24-7 Weight : 0.05025kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
690632-24-7 | |
301.211 | |
QJROOQWDMKQWEZ-UHFFFAOYSA-N | |
2795497 | |
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(S2)C3=CC=CC=C3)C |
C16H20BNO2S | |
MFCD05865106 | |
4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-thiazole, 4-methyl-2-phenyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-thiazole, 4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiazole, 4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1,3-thiazole | |
4-methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole |
Specifications
4-Methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole | |
250mg | |
96.5 | |
97% | |
MFCD05865106 | |
QJROOQWDMKQWEZ-UHFFFAOYSA-N | |
4-methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole | |
2795497 | |
97% |
Amber Glass Bottle | |
690632-24-7 | |
100.0 | |
C16H20BNO2S | |
4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-thiazole, 4-methyl-2-phenyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-thiazole, 4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiazole, 4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1,3-thiazole | |
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(S2)C3=CC=CC=C3)C | |
301.211 | |
301.22 |