missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3522533
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
915707-58-3
C9H10N2O3
194.19
MFCD09065017
JTDUYLQIBHWRRK-UHFFFAOYSA-N
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid, 2h-pyrido 3,2-b-1,4-oxazine-7-carboxylicacid, 3,4-dihydro-4-methyl, 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylicacid, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazine-7-carboxylic acid, 7-carboxy-3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b-1,4-oxazine-7-carboxylic acid, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid
24229645
4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazine-7-carboxylic acid
CN1CCOC2=C1N=CC(=C2)C(=O)O
This item is not returnable. View return policy
915707-58-3
C9H10N2O3
194.19
MFCD09065017
JTDUYLQIBHWRRK-UHFFFAOYSA-N
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid, 2h-pyrido 3,2-b-1,4-oxazine-7-carboxylicacid, 3,4-dihydro-4-methyl, 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylicacid, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazine-7-carboxylic acid, 7-carboxy-3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b-1,4-oxazine-7-carboxylic acid, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid
24229645
4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazine-7-carboxylic acid
CN1CCOC2=C1N=CC(=C2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'