Learn More
4-Methylaminophenol sulfate, ACS reagent, 99.0-101.5%, ACROS Organics™
Brand: Acros Organics 434445000
Code : 29
Additional Details : CAS Number : 55-55-0 Weight : 0.55000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Packaging | Plastic bottle |
---|---|
Quantity | 500g |
Chemical Identifiers
55-55-0 | |
C14H18N2O2·H2SO4 | |
344.38 | |
Metol |
Specifications
4-Methylaminophenol sulfate | |
0.1% max. | |
0.1% max | |
passes | |
260.0°C | |
55-55-0 | |
(CH3NHC6H4OH)2·H2SO4 | |
Metol | |
344.38 | |
Crystalline Powder | |
ACS Reagent |
1.5770g/mL | |
Plastic bottle | |
1.577 | |
White | |
500g | |
C14H18N2O2·H2SO4 | |
15,6091 | |
Solubility in water: 50g/L (20°C). Other solubilities: soluble in 20 parts cold water,soluble in 6 parts boiling water,slightly soluble in alcohol,insoluble in ether | |
344.38 | |
99.0-101.5% |
Safety and Handling
- 4-Methylaminophenol sulfate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Respiratory sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.