missing translation for 'onlineSavingsMsg'
Learn More

4-Methylaminophenol sulfate, ACS reagent, 99.0-101.5%, Thermo Scientific Chemicals

Catalog Number 11975301
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
55-55-0
C14H18N2O2·H2SO4
344.38
Metol
55-55-0
C14H18N2O2·H2SO4
344.38
Metol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 260.0°C
Color White
Density 1.5770g/mL
Ignition Residue 0.1% max.
Packaging Plastic bottle
Linear Formula (CH3NHC6H4OH)2·H2SO4
Quantity 100 g
Merck Index 15,6091
Specific Gravity 1.577
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in 20 parts cold water,soluble in 6 parts boiling water,slightly soluble in alcohol,insoluble in ether
Formula Weight 344.38
Percent Purity 99.0-101.5%
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylaminophenol sulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Methylaminophenol sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'