missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzophenone, 97%, Thermo Scientific Chemicals

Catalog Number 10011430
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
134-84-9
C14H12O
196.25
MFCD00008553
WXPWZZHELZEVPO-UHFFFAOYSA-N
4-methylbenzophenone, phenyl p-tolyl methanone, 4-methylphenyl phenyl methanone, p-methylbenzophenone, methanone, 4-methylphenyl phenyl, p-benzoyltoluene, 4-methyl benzophenone, phenyl p-tolyl ketone, benzophenone, 4-methyl, p-benzophenone, methyl
8652
(4-methylphenyl)-phenylmethanone
CC1=CC=C(C=C1)C(=O)C2=CC=CC=C2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
134-84-9
C14H12O
196.25
MFCD00008553
WXPWZZHELZEVPO-UHFFFAOYSA-N
4-methylbenzophenone, phenyl p-tolyl methanone, 4-methylphenyl phenyl methanone, p-methylbenzophenone, methanone, 4-methylphenyl phenyl, p-benzoyltoluene, 4-methyl benzophenone, phenyl p-tolyl ketone, benzophenone, 4-methyl, p-benzophenone, methyl
8652
(4-methylphenyl)-phenylmethanone
CC1=CC=C(C=C1)C(=O)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 53°C to 57°C
Color Beige to White
Boiling Point 326°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C6H4COC6H5
Quantity 50 g
Beilstein 07, 440
Merck Index 14, 7317
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and chloroform
Formula Weight 196.25
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Methylbenzophenone, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'