missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-n-Butoxy-2,6-difluorobenzonitrile, 97%

Brand:  Alfa Aesar™ H33023.03

Code : NEW

Additional Details : Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15475807

  • 130.00 € / 1g


Estimated Shipment date
from Supplier 09-05-2019
Add to basket

Specifications

Chemical Identifiers

1373920-94-5
211.212
DRMAQHPUVUFLAE-UHFFFAOYSA-N
86277629
CCCCOC1=CC(=C(C(=C1)F)C#N)F
C11H11F2NO
MFCD22201026
4-n-butoxy-2,6-difluorobenzonitrile
4-butoxy-2,6-difluorobenzonitrile

Specifications

4-n-Butoxy-2,6-difluorobenzonitrile
1373920-94-5
MFCD22201026
4-n-butoxy-2,6-difluorobenzonitrile
CCCCOC1=CC(=C(C(=C1)F)C#N)F
211.212
211.21
97%
1g
C11H11F2NO
UN3439
DRMAQHPUVUFLAE-UHFFFAOYSA-N
4-butoxy-2,6-difluorobenzonitrile
86277629
Solid
Videos
SDS